Learn More
Grainger Ball End Mill Single End 2.00mm Carbide
Catalog No. 502751096
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has 2 flutes and a right-hand spiral. It is designed for general purpose milling in materials like aluminum, brass, bronze, and plastics. The end mill features TiAlN coating for increased tool life and heat resistance. It can be run at speeds up to 11,000 RPM. The end mill is made in the USA.
Catalog No. 50-275-1096 Supplier Grainger Supplier No. 14K378
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has 2 flutes and a right-hand spiral. It is designed for general purpose milling in materials like aluminum, brass, bronze, and plastics. The end mill features TiAlN coating for increased tool life and heat resistance. It can be run at speeds up to 11,000 RPM. The end mill is made in the USA.