Learn More
Grainger Ball End Mill Single End 2.00mm Carbide
Catalog No. 502751282
Encompass_Preferred
The Grainger 14K377 is a 14 gauge, type 1 solid copper electrical wire with THHN insulation. It is rated up to 90°C and 600 volts. This wire is UL listed and RoHS compliant. The 14K377 comes in multiple lengths, with the most common being 250 ft reels. This wire is designed for use in conduit and general purpose wiring.
Catalog No. 50-275-1282 Supplier Grainger Supplier No. 14K377
The Grainger 14K377 is a 14 gauge, type 1 solid copper electrical wire with THHN insulation. It is rated up to 90°C and 600 volts. This wire is UL listed and RoHS compliant. The 14K377 comes in multiple lengths, with the most common being 250 ft reels. This wire is designed for use in conduit and general purpose wiring.