Learn More
Grainger Ball End Mill Single End 4.00mm Carbide
Catalog No. 502751816
Encompass_Preferred
The Grainger 14K741 is a 14 gauge, type 1 stainless steel wire rope with 7x19 construction. It has an independent wire rope core and is right regular lay. This wire rope is uncoated and non-rotating. It has improved fatigue resistance and is designed for general purpose use. This wire rope meets federal specification RR-W-410 and military specification MIL-W-83420. It comes in multiple lengths up to 500 ft and has a breaking strength of 15,400 lbs.
Catalog No. 50-275-1816 Supplier Grainger Supplier No. 14K741
The Grainger 14K741 is a 14 gauge, type 1 stainless steel wire rope with 7x19 construction. It has an independent wire rope core and is right regular lay. This wire rope is uncoated and non-rotating. It has improved fatigue resistance and is designed for general purpose use. This wire rope meets federal specification RR-W-410 and military specification MIL-W-83420. It comes in multiple lengths up to 500 ft and has a breaking strength of 15,400 lbs.