Learn More
Grainger Ball End Mill Single End 4.00mm Carbide
Catalog No. 502749744
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 1/2" cutting diameter, a 2" overall length, and a 2 flute design. This end mill is center cutting and right hand spiral. It can be used for slotting, profiling, contouring, and facing on materials like steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. The ball nose design allows milling of 3D contoured shapes.
Catalog No. 50-274-9744 Supplier Grainger Supplier No. 14K745
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/2" shank diameter, a 1/2" cutting diameter, a 2" overall length, and a 2 flute design. This end mill is center cutting and right hand spiral. It can be used for slotting, profiling, contouring, and facing on materials like steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic, and wood. The ball nose design allows milling of 3D contoured shapes.