Learn More
Grainger Ball End Mill Single End 4.00mm Carbide
Catalog No. 502751500
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball end for milling contoured surfaces. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is made of M42 high-speed steel for wear resistance and has TiAlN coating for heat resistance and lubricity. The end mill can be run at speeds up to 325 SFM and is well-suited for slotting, side milling and profiling operations.
Catalog No. 50-275-1500 Supplier Grainger Supplier No. 14K744
The Ball End Mill from Ball is a high-speed steel end mill with a ball end for milling contoured surfaces. It has a 1/2" shank diameter, a 2" overall length, and a 1/2" cutting diameter. The end mill has 4 flutes and a right-hand spiral. It is made of M42 high-speed steel for wear resistance and has TiAlN coating for heat resistance and lubricity. The end mill can be run at speeds up to 325 SFM and is well-suited for slotting, side milling and profiling operations.