Learn More
Grainger Ball End Mill Single End 4.00mm Carbide
Catalog No. 502751760
Encompass_Preferred
The Grainger 14K757 is a 14-gauge, 7-strand galvanized steel aircraft cable with a breaking strength of 5,400 lbs. It has a diameter of 0.162 inches and is sold by the foot, with a minimum order length of 50 feet. The cable is very flexible and is commonly used for industrial lifting applications. It features 7x19 strand construction and is manufactured in the USA. The galvanized coating provides good corrosion resistance. This aircraft cable meets military specification MIL-W-83420.
Catalog No. 50-275-1760 Supplier Grainger Supplier No. 14K757
The Grainger 14K757 is a 14-gauge, 7-strand galvanized steel aircraft cable with a breaking strength of 5,400 lbs. It has a diameter of 0.162 inches and is sold by the foot, with a minimum order length of 50 feet. The cable is very flexible and is commonly used for industrial lifting applications. It features 7x19 strand construction and is manufactured in the USA. The galvanized coating provides good corrosion resistance. This aircraft cable meets military specification MIL-W-83420.