Learn More
Grainger Approved Vendor Ball End Mill, Carbide, AlTiN Coated, 2 Flute, 5/32 in., 1 ea.
Catalog No. 502783294
Encompass_Preferred
This solid carbide ball nose end mill is designed for general purpose machining applications, offering superior wear and heat resistance. It features two flutes and a center-cutting design, making it suitable for plunging, ramping, and milling complex shapes with round-bottomed grooves and slots. The AlTiN coating provides high heat resistance, with an oxidation temperature of 800°C and a hardness of 4500 HV, optimizing it for milling abrasive materials.
  • Carbide construction for increased durability and heat resistance
  • AlTiN coating for performance in hard and abrasive materials
  • Center-cutting design for plunge cutting and bottom surface machining
  • 2-flute configuration with a 30° helix angle
  • Single end design with a 3/16 in. Weldon flat shank
  • 5/32 in. milling diameter with 9/16 in. length of cut and 2 in. overall length
Catalog No. 50-278-3294 Supplier Grainger Supplier No. 19LN39
This solid carbide ball nose end mill is designed for general purpose machining applications, offering superior wear and heat resistance. It features two flutes and a center-cutting design, making it suitable for plunging, ramping, and milling complex shapes with round-bottomed grooves and slots. The AlTiN coating provides high heat resistance, with an oxidation temperature of 800°C and a hardness of 4500 HV, optimizing it for milling abrasive materials.
  • Carbide construction for increased durability and heat resistance
  • AlTiN coating for performance in hard and abrasive materials
  • Center-cutting design for plunge cutting and bottom surface machining
  • 2-flute configuration with a 30° helix angle
  • Single end design with a 3/16 in. Weldon flat shank
  • 5/32 in. milling diameter with 9/16 in. length of cut and 2 in. overall length