Learn More
Grainger Ball End Mill Single End 6.00mm Carbide
Catalog No. 502751273
Encompass_Preferred
The Micro 100 Ball End Mill is a 4-flute carbide end mill with a ball end. It has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2". The ball end allows milling of 3D contoured surfaces. It can machine steels, stainless steels, cast iron, non-ferrous metals, and non-metallic materials. The 4-flute design provides good chip evacuation. This end mill is made of solid carbide for wear resistance and has TiAlN coating for heat resistance and lubricity. It has center-cutting geometry allowing plunge cuts into material. Recommended speeds are 20,000-30,000 RPM with feed rates of 8-20 IPM.
Catalog No. 50-275-1273 Supplier Grainger Supplier No. 14K770
The Micro 100 Ball End Mill is a 4-flute carbide end mill with a ball end. It has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2". The ball end allows milling of 3D contoured surfaces. It can machine steels, stainless steels, cast iron, non-ferrous metals, and non-metallic materials. The 4-flute design provides good chip evacuation. This end mill is made of solid carbide for wear resistance and has TiAlN coating for heat resistance and lubricity. It has center-cutting geometry allowing plunge cuts into material. Recommended speeds are 20,000-30,000 RPM with feed rates of 8-20 IPM.