Learn More
Grainger Ball End Mill Single End 6.00mm Carbide
Catalog No. 502753298
Encompass_Preferred
The Micro 100 Ball End Mill is a 2 flute carbide end mill with a ball end. It has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2". The ball end allows milling of 3D contoured surfaces. Carbide provides wear resistance and tool life. It can machine steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. The 2 flute design provides efficient chip ejection. Made in USA by Micro 100, a manufacturer of high performance cutting tools.
Catalog No. 50-275-3298 Supplier Grainger Supplier No. 14M595
The Micro 100 Ball End Mill is a 2 flute carbide end mill with a ball end. It has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2". The ball end allows milling of 3D contoured surfaces. Carbide provides wear resistance and tool life. It can machine steels, stainless steels, cast iron, aluminum, and other non-ferrous materials. The 2 flute design provides efficient chip ejection. Made in USA by Micro 100, a manufacturer of high performance cutting tools.