Learn More
Grainger Ball End Mill Single End 6.00mm Carbide
Catalog No. 502749581
Encompass_Preferred
The SGS TOOL Ball End Mill Carbide is a 4 flute ball end mill made of solid carbide with a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. It has a TiAIN coating and is designed for general purpose milling in steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. The 4 flute design provides good chip ejection and finish. This is a high quality carbide end mill made in Germany suitable for high speed machining in a range of materials.
Catalog No. 50-274-9581 Supplier Grainger Supplier No. 14K775
The SGS TOOL Ball End Mill Carbide is a 4 flute ball end mill made of solid carbide with a 1/4" shank diameter, 1/2" cutting diameter, and 2" overall length. It has a TiAIN coating and is designed for general purpose milling in steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. The 4 flute design provides good chip ejection and finish. This is a high quality carbide end mill made in Germany suitable for high speed machining in a range of materials.