Learn More
Grainger Ball End Mill Single End 6.00mm Carbide
Catalog No. 502750351
Encompass_Preferred
The Grainger 6mm carbide ball end mill is a single end milling cutter with a 6mm diameter ball nose end. It has 2 flutes and is made of solid carbide for wear resistance when milling steel, hardened steel, cast iron, aluminum, copper, brass and other non-ferrous metals. This end mill has a right hand spiral flute for clockwise milling and features TiAlN coating for heat and wear resistance. It has an overall length of 50mm and shank diameter of 6mm. The ball end mill is designed for high speed machining in both radial and axial directions.
Catalog No. 50-275-0351 Supplier Grainger Supplier No. 14K762
The Grainger 6mm carbide ball end mill is a single end milling cutter with a 6mm diameter ball nose end. It has 2 flutes and is made of solid carbide for wear resistance when milling steel, hardened steel, cast iron, aluminum, copper, brass and other non-ferrous metals. This end mill has a right hand spiral flute for clockwise milling and features TiAlN coating for heat and wear resistance. It has an overall length of 50mm and shank diameter of 6mm. The ball end mill is designed for high speed machining in both radial and axial directions.