Learn More
Grainger Ball End Mill Single End 8.00mm Carbide
Catalog No. 502749504
Encompass_Preferred
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, 1/2" cutting diameter, 2" overall length, and 2 flute design. This end mill is center cutting and suitable for slotting, side milling and profiling operations. It can machine steels, stainless steels, cast iron, non-ferrous metals and non-metallic materials. The ball nose geometry provides smooth milling action and accurate tracing of 3D contours.
Catalog No. 50-274-9504 Supplier Grainger Supplier No. 14K788
The Ball End Mill from Ball is a high-speed steel end mill with a ball nose end. It has a 1/4" shank diameter, 1/2" cutting diameter, 2" overall length, and 2 flute design. This end mill is center cutting and suitable for slotting, side milling and profiling operations. It can machine steels, stainless steels, cast iron, non-ferrous metals and non-metallic materials. The ball nose geometry provides smooth milling action and accurate tracing of 3D contours.