Learn More
Hach Company Ball Float Mount Kit
Supplier:  Hach Company 5794300
Encompass
Ball Float Mount Kit for the Hach LDO Dissolved Oxygen Probe PVC swivel assembly inflatable ball float assembly and 2 4m (8 ft) PVC pole
Catalog No. 50-002-07175