Learn More
Grainger Ball Foot Air Chuck
Catalog No. 502750748
Encompass_Preferred
The Legacy Clip-on Ball Foot Air Chuck is a pneumatic air chuck that attaches to air hoses to inflate tires and other inflatables. It has a 1/4" NPT thread to connect to the air hose. The chuck has a rubber-coated ball foot that seals against the inflatable's valve stem when pressed down. It is made of brass and has a max pressure rating of 300 PSI. The Legacy air chuck provides a hands-free way to inflate tires and equipment.
Catalog No. 50-275-0748 Supplier Grainger Supplier No. 140W96
The Legacy Clip-on Ball Foot Air Chuck is a pneumatic air chuck that attaches to air hoses to inflate tires and other inflatables. It has a 1/4" NPT thread to connect to the air hose. The chuck has a rubber-coated ball foot that seals against the inflatable's valve stem when pressed down. It is made of brass and has a max pressure rating of 300 PSI. The Legacy air chuck provides a hands-free way to inflate tires and equipment.