Learn More
World Precision Instrument Ball Joint, 4 cm long
SDP
Supplier:  World Precision Instrument 500476
Encompass
A Ball Joint Attachment Arm for a micromanipulator gives you flexibility and precise positioning options. With a ball joint attachment you can adjust the orientation and angle of the micromanipulator's probe. The ball joint allows for a 360º range of movement, including rotational movement and gives you greater maneuverability. Now you can comfortably position your micro manipulator to suit you, reducing physical strain and fatigue. The Ball Joint Attachment Arm provides a versatile connection point so you can easily change the probe or handle. Choose between the 7 cm (2.76") or the 4 cm (1.58") arm. Choose the longer arm for probe holders with a diameter of 5-9 mm or the shorter arm for smaller electrode holders with a diameter of 2.8-4.5 mm. These micro manipulator attachment arms work with the MMJ Joystick Controlled Manual Manipulator, the M325 Micrometer Slide Micromanipulator, the MD4 Dual Tool Holder Micromanipulator, or the KITE Economy Micromanipulator.
Catalog No. 50-252-1443