Learn More
Thermo Scientific BALL JOINT CLIP
Catalog No. NC0193912 Shop All Thermo Scientific Products
Encompass
Ball Joint Clip; Metal ball joint clip for connecting spraychamber and adaptor.; Compatible with all iCAP 6000 Series Spectrometers.
Catalog No. NC0193912 Supplier Thermo Scientific Supplier No. 842312051421
Ball Joint Clip; Metal ball joint clip for connecting spraychamber and adaptor.; Compatible with all iCAP 6000 Series Spectrometers.