Learn More
Thermo Scientific Chemicals Ball pestle; clear plexiglas; 1/8in. diameter
Catalog No. AA44532KN
Change view
Click to view available options
Quantity:
100/Pk.
25/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44532KN 100/Pk.
AA44532KM 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44532KN Supplier Thermo Scientific Chemicals Supplier No. 044532KN
Add to Cart
For use with Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 0.125 in.
Diameter (Metric) Outer 3.17mm
Product Type Ball Pestle
For Use With (Equipment) Wig-L-Bug Mixing/Grinding Mill (AA44204KT and AA44604KT)
Quantity 100/Pk.