Learn More
Chemplex Industries BALL PESTLES PLASTIC 7MM
Catalog No. 50249458
Encompass_Preferred
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Methylmethacrylate; Plastic; 7mm
Catalog No. 50-249-458 Supplier Chemplex Industries Supplier No. 1207
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Methylmethacrylate; Plastic; 7mm