Learn More
Chemplex Industries BALL PESTLES PLASTIC 8MM
Catalog No. 50249459
Encompass_Preferred
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Methylmethacrylate; Plastic; 8mm
Catalog No. 50-249-459 Supplier Chemplex Industries Supplier No. 1208
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Methylmethacrylate; Plastic; 8mm