Learn More
MSC Midwest Control Miniature Ball Valve, 1/2 in. FNPT, Brass, 450 PSI, 1 ea.
Catalog No. 501909772
Encompass
This 1/2 in. FPT miniature ball valve features a one-piece nickel-plated brass body designed for reliable on-off service. Suitable for vacuum service up to 29.9 in. Hg, this valve is rated for 450 PSI CWP and 150 PSI WSP. It includes a chrome-plated brass ball, PTFE ball seats, and a Viton™ O-ring, with a design that supports bi-directional operation and panel mounting.
  • One-piece nickel-plated brass body with blow-out proof stem
  • Chrome-plated brass ball with PTFE seats and Viton™ O-ring
  • Rated for 450 PSI CWP and 150 PSI WSP
  • Suitable for vacuum service up to 29.9 in. Hg
  • Supports bi-directional flow
  • Panel-mountable configuration
  • Includes interchangeable wedge, tee, and lever handles
  • Meets MSS SP-110 standards and is RoHS compliant
Catalog No. 50-190-9772 Supplier MSC Supplier No. 89495196
This 1/2 in. FPT miniature ball valve features a one-piece nickel-plated brass body designed for reliable on-off service. Suitable for vacuum service up to 29.9 in. Hg, this valve is rated for 450 PSI CWP and 150 PSI WSP. It includes a chrome-plated brass ball, PTFE ball seats, and a Viton™ O-ring, with a design that supports bi-directional operation and panel mounting.
  • One-piece nickel-plated brass body with blow-out proof stem
  • Chrome-plated brass ball with PTFE seats and Viton™ O-ring
  • Rated for 450 PSI CWP and 150 PSI WSP
  • Suitable for vacuum service up to 29.9 in. Hg
  • Supports bi-directional flow
  • Panel-mountable configuration
  • Includes interchangeable wedge, tee, and lever handles
  • Meets MSS SP-110 standards and is RoHS compliant