Learn More
Ace Glass, Inc. BALL VALVE,2in NPT,SS,PTFE
SDP
Catalog No. 5000854802
Encompass
On/Off ball valve with lockable handle (padlock not included). Wetted surfaces are stainless steel and PTFE.
Catalog No. 50-008-54802 Supplier Ace Glass, Inc. Supplier No. 1382401
On/Off ball valve with lockable handle (padlock not included). Wetted surfaces are stainless steel and PTFE.