Learn More
Grainger Speedaire Ball Valve, Full Port, Brass, Lever Handle, 1 Ea.
Catalog No. 502779124
Encompass_Preferred
This brass ball valve functions as a reliable on/off shut-off valve designed for industrial pneumatic applications and air compressor systems. It serves as a replacement component to repair worn or damaged parts in compatible air compressor equipment.
  • Brass construction for durability
  • Full port design
  • Lever handle for easy manual operation
  • Designed for use in air compressor systems
Catalog No. 50-277-9124 Supplier Grainger Supplier No. 19NC01
This brass ball valve functions as a reliable on/off shut-off valve designed for industrial pneumatic applications and air compressor systems. It serves as a replacement component to repair worn or damaged parts in compatible air compressor equipment.
  • Brass construction for durability
  • Full port design
  • Lever handle for easy manual operation
  • Designed for use in air compressor systems