Learn More
Grainger Ballcock Connectr 1/2 CTS x7/8-15/16 UNS
Catalog No. 502765957
Encompass_Preferred
The Grainger Approved Ballcock Connector is a 1/2" CTS x 7/8"-15/16" UNS brass ballcock connector. It is made of chrome-plated brass and has a maximum pressure rating of 125 psi and maximum temperature rating of 250°F. This connector has a swivel nut for easy installation and is designed to connect a ballcock fill valve to a water supply line in a toilet tank. It measures approximately 2-1/2" in overall length. The Grainger Approved brand offers quality plumbing parts at an economical price point. This ballcock connector provides a reliable connection for refilling the toilet tank after flushing.
Catalog No. 50-276-5957 Supplier Grainger Supplier No. 16T757
The Grainger Approved Ballcock Connector is a 1/2" CTS x 7/8"-15/16" UNS brass ballcock connector. It is made of chrome-plated brass and has a maximum pressure rating of 125 psi and maximum temperature rating of 250°F. This connector has a swivel nut for easy installation and is designed to connect a ballcock fill valve to a water supply line in a toilet tank. It measures approximately 2-1/2" in overall length. The Grainger Approved brand offers quality plumbing parts at an economical price point. This ballcock connector provides a reliable connection for refilling the toilet tank after flushing.