Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Ballistics Car
GREENER_CHOICESCI_ED

Catalog No. S139426 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139426 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139426 Supplier Supertek™ Supplier No. PH10537
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Ballistics Car demonstrates that the forward motion of a ball ejected vertically is the same as the vehical from which it was ejected by.

  • Comes with 1 car with ball bearing wheels and vertically mounted spring loaded barrel.
  • The barrel permits two spring tensions.
  • Complete with a 25 mm steel ball and release pin.
  • Instructions included

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Force and Mass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.