Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Balloon Puck Set
GREENER_CHOICESCI_ED

Catalog No. S139424 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139424 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139424 Supplier Supertek™ Supplier No. PH10516
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Supertek™ Balloon Puck Set explores the laws of motion without an air table and studies the inertia, constant velocity and acceleration, and collisions in two dimensions.

  • On a flat smooth surface, the balloon puck can glide through without much resistance.
  • Pucks ride on a film of air produced by air escaping from the balloons, which are inflated and attached to the pucks.
  • Kit Includes...
  • 2 large pucks (5.51 in. dia.)
  • 2 small pucks (4 in. dia)
  • 4 rubber stoppers
  • 4 tubes
  • 4 balloons
  • 4 rubber bands.
  • Instructions included

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.