Learn More
Chemplex Industries BALLPESTLES TUNGCARBIDE 7MM
Catalog No. 50249462
Encompass_Preferred
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Tungsten Carbide; 6 units per package, 7mm
Catalog No. 50-249-462 Supplier Chemplex Industries Supplier No. 1236
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Tungsten Carbide; 6 units per package, 7mm