Learn More
Chemplex Industries BALLPESTLES TUNGCARBIDE 8MM
Catalog No. 50249463
Encompass_Preferred
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Tungsten Carbide; 6 units per package, 8mm
Catalog No. 50-249-463 Supplier Chemplex Industries Supplier No. 1237
CHEMPLEX BALL PESTLES: FOR SPECTROVIALS(TM) Comminution Vials, Tungsten Carbide; 6 units per package, 8mm