Learn More
MSC Bally Block Co. Locker Bench, Solid Maple, 48 x 9.5 in
Catalog No. 5000531935
Encompass
This locker bench is constructed from solid northern hard maple, offering durability and a natural aesthetic. It features a Durakryl 102 UV-cured urethane finish, providing superior moisture impedance. The bench measures 48 inches long, 9.5 inches wide, and 1.25 inches thick, suitable for institutional and industrial environments.
  • Constructed from solid northern hard maple.
  • Features a Durakryl 102 UV-cured urethane finish for moisture impedance.
  • Dimensions of 48 in long, 9.5 in wide, and 1.25 in thick.
  • Edges and corners have a 3/8 in radius.
  • Made with a strong and flexible PVA adhesive for high durability.
Catalog No. 50-005-31935 Supplier MSC Supplier No. 89765903
This locker bench is constructed from solid northern hard maple, offering durability and a natural aesthetic. It features a Durakryl 102 UV-cured urethane finish, providing superior moisture impedance. The bench measures 48 inches long, 9.5 inches wide, and 1.25 inches thick, suitable for institutional and industrial environments.
  • Constructed from solid northern hard maple.
  • Features a Durakryl 102 UV-cured urethane finish for moisture impedance.
  • Dimensions of 48 in long, 9.5 in wide, and 1.25 in thick.
  • Edges and corners have a 3/8 in radius.
  • Made with a strong and flexible PVA adhesive for high durability.