Learn More
MSC Ballymore Rolling Ladder, Steel, 2 Step, 19 in Platform Height
Catalog No. 5000514964
Encompass
A spring-loaded rolling ladder constructed from gray, powder-coated steel. It has a 19 in overall height, a 19 in platform height, and two steps. The ladder features a 16 in platform width, a 20 in base width, and perforated treads. It is equipped with spring-loaded swivel casters that retract under user weight and meets OSHA and ANSI standards. Assembly is required.
  • Constructed from steel
  • Two steps
  • 19 in platform height
  • 16 in platform width
  • 20 in base width
  • Perforated treads
  • Gray, powder-coated finish
  • Spring-loaded swivel casters retract under user weight
  • Meets OSHA and ANSI standards
  • Assembly is required
Catalog No. 50-005-14964 Supplier MSC Supplier No. 80591142
A spring-loaded rolling ladder constructed from gray, powder-coated steel. It has a 19 in overall height, a 19 in platform height, and two steps. The ladder features a 16 in platform width, a 20 in base width, and perforated treads. It is equipped with spring-loaded swivel casters that retract under user weight and meets OSHA and ANSI standards. Assembly is required.
  • Constructed from steel
  • Two steps
  • 19 in platform height
  • 16 in platform width
  • 20 in base width
  • Perforated treads
  • Gray, powder-coated finish
  • Spring-loaded swivel casters retract under user weight
  • Meets OSHA and ANSI standards
  • Assembly is required