Learn More
Medchemexpress LLC Balofloxacin | 127294-70-6 | 98.3% | 250 MG
SDP
Catalog No. 5000406162
Encompass_Preferred
Balofloxacin is an orally active fluoroquinolone antibiotic that exhibits broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria. It can be used for the research of respiratory, intestinal, and urinary tract infections.
  • Orally active fluoroquinolone antibiotic
  • Broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria
  • Can be used for research of respiratory, intestinal, and urinary tract infections
Catalog No. 50-004-06162 Supplier Medchemexpress LLC Supplier No. 250MGHYB0159
Balofloxacin is an orally active fluoroquinolone antibiotic that exhibits broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria. It can be used for the research of respiratory, intestinal, and urinary tract infections.
  • Orally active fluoroquinolone antibiotic
  • Broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria
  • Can be used for research of respiratory, intestinal, and urinary tract infections