Learn More
AdipoGen Balofloxacin
SDP
Supplier:  AdipoGen CDXB0302M100
Encompass_Preferred
Chemical. CAS 127294-70-6. Formula C20H24FN3O4. MW 389.42. Synthetic. Fluoroquinolone antibiotic with potent bactericidal activity. Inhibits the synthesis of bacterial DNA by interfering with the the supercoiling activity of DNA gyrase.
Catalog No. 50-256-1600