Learn More
Medchemexpress LLC HY-B0159 500mg Medchemexpress, Balofloxacin CAS:127294-70-6 Purity:>98%
SDP
Catalog No. 502082385
Encompass_Preferred
Medchemexpress, HY-B0159 500mg Balofloxacin CAS:127294-70-6 Balofloxacin (Q-35) is an orally active fluoroquinolone antibiotic with broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria. Balofloxacin can be used for the research of respiratory, intestinal, and urinary tract infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2385 Supplier Medchemexpress LLC Supplier No. HYB0159500MG
Medchemexpress, HY-B0159 500mg Balofloxacin CAS:127294-70-6 Balofloxacin (Q-35) is an orally active fluoroquinolone antibiotic with broad-spectrum antibacterial activity against gram-negative, gram-positive, and anaerobic bacteria. Balofloxacin can be used for the research of respiratory, intestinal, and urinary tract infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.