Learn More
Medchemexpress LLC HY-109024 5mg Medchemexpress, Balovaptan CAS:1228088-30-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090245MG
Encompass_Preferred
Medchemexpress, HY-109024 5mg Balovaptan CAS:1228088-30-9 Balovaptan (RG7314) is a highly potent and selective brain-penetrant vasopressin 1a (hV1a) receptor antagonist, with Kis of 1 and 39 nM for human (hV1a) and mouse (mV1a) receptors, and is used for the research of autism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8651