Learn More
Medchemexpress LLC HY-109025A 10mg Medchemexpress, Baloxavir CAS:1985605-59-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY109025A10MG
Encompass_Preferred
Medchemexpress, HY-109025A 10mg Baloxavir CAS:1985605-59-1 Baloxavir (Baloxavir acid) is a first-in-class, potent and selective cap-dependent endonuclease (CEN) inhibitor within the polymerase PA subunit of influenza A and B viruses. Baloxavir inhibits viral RNA transcription and replication and has potently antiviral activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8789