Learn More
Medchemexpress LLC Baloxavir acid-d4 | 2415027-80-2 | 99.3% | 487.51 | C24H15D4F2N3O4S | 1mg
SDP
Catalog No. 5000269364
Encompass_Preferred
Baloxavir-d4 (Baloxavir acid-d4 S-033447-d4) is the deuterium-labeled Baloxavir (HY-109025A) Baloxavir-d4 (Baloxavir-d4 acid) derived from the proagent Baloxavir-d4 marboxil is a first-in-class potent and selective cap-dependent endonuclease (CEN) inhibitor within the polymerase PA subunit of influenza A and B viruses Baloxavir-d4 inhibits viral RNA transcription and replication and has potently antiviral activity[1][2]
Catalog No. 50-002-69364 Supplier Medchemexpress LLC Supplier No. HYW7518351MG
Baloxavir-d4 (Baloxavir acid-d4 S-033447-d4) is the deuterium-labeled Baloxavir (HY-109025A) Baloxavir-d4 (Baloxavir-d4 acid) derived from the proagent Baloxavir-d4 marboxil is a first-in-class potent and selective cap-dependent endonuclease (CEN) inhibitor within the polymerase PA subunit of influenza A and B viruses Baloxavir-d4 inhibits viral RNA transcription and replication and has potently antiviral activity[1][2]