Learn More
Apexbio Technology LLC Baloxavir marboxil, 10mg CAS# 1985606-14-1
SDP
Supplier:  Apexbio Technology LLC B845310
Encompass_Preferred
Baloxavir marboxil is an antiviral drug and is an endonuclease inhibitor. In rats and monkeys, the blood concentration of the drug is lower than the minimum detection amount in a single oral administration of the drug. In preclinical influenza A and B infection models (including strains that are resistant to existing antiviral drugs), Baloxavir marboxil has a certain effect. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8458