Learn More
Medchemexpress LLC Baloxavir marboxil | 1985606-14-1 | 99.9% | 571.55 | 250MG
SDP
Catalog No. 5000347547
Encompass_Preferred
Baloxavir marboxil is a selective inhibitor of influenza cap-dependent endonuclease. It is a potent antiviral agent that shows activity against influenza A and B virus.
  • Selective inhibitor of influenza cap-dependent endonuclease
  • Potent antiviral agent
  • Active against influenza A and B virus
Catalog No. 50-003-47547 Supplier Medchemexpress LLC Supplier No. 250MGHY109025
Baloxavir marboxil is a selective inhibitor of influenza cap-dependent endonuclease. It is a potent antiviral agent that shows activity against influenza A and B virus.
  • Selective inhibitor of influenza cap-dependent endonuclease
  • Potent antiviral agent
  • Active against influenza A and B virus