Learn More
TARGETMOL CHEMICALS INC BALOXAVIR MARBOXIL 250MG
SDP
Catalog No. 502579494
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Baloxavir marboxil (S-033188) is a novel cap-dependent endonuclease inhibitor for the treatment of influenza A and B virus infections. purity: 98%
Catalog No. 50-257-9494 Supplier TARGETMOL CHEMICALS INC Supplier No. T6195250MG
Also available in 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Baloxavir marboxil (S-033188) is a novel cap-dependent endonuclease inhibitor for the treatment of influenza A and B virus infections. purity: 98%