Learn More
Medchemexpress LLC HY-B0667 10mg Medchemexpress, Balsalazide CAS:80573-04-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB066710MG
Encompass_Preferred
Medchemexpress, HY-B0667 10mg Balsalazide CAS:80573-04-2 Balsalazide could suppress colitis-associated carcinogenesis through modulation of IL-6/STAT3 pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2637