Learn More
Cayman Chemical Balsalazide, Purity: ≥98%, CAS: 80573-04-2, 50 mg
SDP
Catalog No. 502980077
Encompass_Preferred
A prodrug form of 5-ASA; cleaved by bacterial azoreductases in the intestinal lumen to release 5-ASA; decreases IL-2 levels and increases IL-6 levels in the serum and colonic mucosa membrane, as well as decreases micro- and macroscopic colonic damage, in a rat model of DNCB-induced colitis at 600 mg/kg per day
Catalog No. 50-298-0077 Supplier Cayman Chemical Supplier No. 1868050 MG
A prodrug form of 5-ASA; cleaved by bacterial azoreductases in the intestinal lumen to release 5-ASA; decreases IL-2 levels and increases IL-6 levels in the serum and colonic mucosa membrane, as well as decreases micro- and macroscopic colonic damage, in a rat model of DNCB-induced colitis at 600 mg/kg per day