Learn More
Cayman Chemical Balsalazide-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502939922
Encompass_Preferred
An internal standard for the quantification of balsalazide by GC- or LC-MS
Catalog No. 50-293-9922 Supplier Cayman Chemical Supplier No. 288881 MG
An internal standard for the quantification of balsalazide by GC- or LC-MS