Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Balsalazide-d4 | 2714315-25-8 | 361.34 | C17H11D4N3O6 | 10 MG
SDP

Catalog No. 5000298188
Encompass_Preferred

Balsalazide-d4 is a deuterium-labeled analogue of balsalazide intended for use as an analytical/internal standard and research reagent in biochemical and pharmacological studies, particularly those involving interleukin and STAT3 signaling. It is supplied as a 10 mg solid with documented storage and solubility information to support analytical workflows.

  • Deuterium-labeled isotopic standard for analytical assays
  • Provided as 10 mg of solid suitable for LC-MS and quantitative analysis
  • Molecular formula C17H11D4N3O6; molecular weight 361.34
  • Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required
  • Storage guidance: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months)
  • Suitable for metabolic tracing, quantitation, and mechanism-of-action studies

Catalog No. 50-002-98188 Supplier Medchemexpress LLC Supplier No. HYB0667S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Balsalazide-d4 is a deuterium-labeled analogue of balsalazide intended for use as an analytical/internal standard and research reagent in biochemical and pharmacological studies, particularly those involving interleukin and STAT3 signaling. It is supplied as a 10 mg solid with documented storage and solubility information to support analytical workflows.

  • Deuterium-labeled isotopic standard for analytical assays
  • Provided as 10 mg of solid suitable for LC-MS and quantitative analysis
  • Molecular formula C17H11D4N3O6; molecular weight 361.34
  • Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required
  • Storage guidance: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months)
  • Suitable for metabolic tracing, quantitation, and mechanism-of-action studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.