Learn More
Medchemexpress LLC Balsalazide-d4 | 2714315-25-8 | 99.9% | 1 MG
SDP
Catalog No. 5000408259
Encompass_Preferred
Balsalazide-d4 is a deuterium-labeled form of Balsalazide. It can be used as a tracer and as an internal standard for quantitative analysis. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suppresses colitis-associated carcinogenesis through modulation of the IL-6/STAT3 pathway
Catalog No. 50-004-08259 Supplier Medchemexpress LLC Supplier No. HYB0667S11MG
Balsalazide-d4 is a deuterium-labeled form of Balsalazide. It can be used as a tracer and as an internal standard for quantitative analysis. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suppresses colitis-associated carcinogenesis through modulation of the IL-6/STAT3 pathway