Learn More
Medchemexpress LLC Balsalazide-d4 | 2714315-25-8 | 99.9% | 5 MG
SDP
Catalog No. 5000408514
Encompass_Preferred
Balsalazide-d4 is deuterium labeled Balsalazide. It can suppress colitis-associated carcinogenesis by modulating the IL-6/STAT3 pathway. This compound is for research use only and not sold to patients. Stable heavy isotopes like deuterium are incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08514 Supplier Medchemexpress LLC Supplier No. HYB0667S15MG
Balsalazide-d4 is deuterium labeled Balsalazide. It can suppress colitis-associated carcinogenesis by modulating the IL-6/STAT3 pathway. This compound is for research use only and not sold to patients. Stable heavy isotopes like deuterium are incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.