Learn More
Selleck Chemical LLC Balsalazide disodium S3745-25mg
SDP
Catalog No. 503110550
Encompass_Preferred
Balsalazide Disodium is the disodium salt form of balsalazide an aminosalicylate and oral prodrug that is enzymatically cleaved in the colon to produce the anti-inflammatory agent mesalazine Balsalazide Disodium suppress colitis-associated carcinogenesis through modulation of IL-6/STAT3 pathway
Catalog No. 50-311-0550 Supplier Selleck Chemical LLC Supplier No. S374525MG
Balsalazide Disodium is the disodium salt form of balsalazide an aminosalicylate and oral prodrug that is enzymatically cleaved in the colon to produce the anti-inflammatory agent mesalazine Balsalazide Disodium suppress colitis-associated carcinogenesis through modulation of IL-6/STAT3 pathway