Learn More
Apexbio Technology LLC Balsalazide-disodium-diH2O
SDP
Catalog No. 5000569698
Encompass_Preferred
Balsalazide disodium dihydrate (CAS No 150399-21-6) is a prodrug commonly used in biomedical research focused on gastrointestinal conditions like ulcerative colitis Originating from efforts to reduce inflammation its mechanism involves releasing the active 5-aminosalicylic acid (5-ASA) in the colon which inhibits the production of inflammatory mediators such as leukotrienes and prostaglandins In vitro it shows activity in the nanomolar to low micromolar range indicating its efficacy in modulating inflammatory pathways Typical applications include studies using cell models like colon epithelial cells and animal studies aimed at understanding its therapeutic potential It is also explored in drug screening for new anti-inflammatory agents Experimental concentrations typically depend on the specific research design allowing flexibility in dosage depending on the experimental setup and objective
Catalog No. 50-005-69698 Supplier Apexbio Technology LLC Supplier No. C645910
Balsalazide disodium dihydrate (CAS No 150399-21-6) is a prodrug commonly used in biomedical research focused on gastrointestinal conditions like ulcerative colitis Originating from efforts to reduce inflammation its mechanism involves releasing the active 5-aminosalicylic acid (5-ASA) in the colon which inhibits the production of inflammatory mediators such as leukotrienes and prostaglandins In vitro it shows activity in the nanomolar to low micromolar range indicating its efficacy in modulating inflammatory pathways Typical applications include studies using cell models like colon epithelial cells and animal studies aimed at understanding its therapeutic potential It is also explored in drug screening for new anti-inflammatory agents Experimental concentrations typically depend on the specific research design allowing flexibility in dosage depending on the experimental setup and objective