Learn More
Perkin Elmer US LLC Balston Type 95A Acetylene Filter
Supplier:  Perkin Elmer US LLC N9301399
Encompass_Preferred
Removes particulate and acetone droplets from acetylene to 99.99% at 0.1 micron protecting your microcomputer gas controls and AA burner system from contamination and corrosion.Recommended for use with AA spectrometers where the air/acetylene filter is not installed. Includes mounting bracket and appropriate fittings for installation on your acetylene line.
Catalog No. NC1898461