Learn More
CPC Scientific H-Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val-Gly-Arg-Pro-Glu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific BAMP001B
Encompass
The precursor of bovine adrenomedullary Met-enkephalin .ONE-LETTER SEQUENCE: YGGFMRRVGRPEMOLECULAR FORMULA: C62H97N21O16S1MOLECULAR WEIGHT:1424.65STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [75513-71-2], RESEARCH AREA: PainREFERENCES: K.Mizuno et al., Biochem. Biophys. Res. Commun., 97, 1283 (1980)
Catalog No. 50-210-8840