Learn More
Medchemexpress LLC HY-110284 10mg Medchemexpress, BAM 15 CAS:210302-17-3 Purity:>98%
SDP
Catalog No. 501968787
Encompass_Preferred
Medchemexpress, HY-110284 10mg BAM 15 CAS:210302-17-3 BAM 15 is a novel mitochondrial protonophore uncoupler. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8787 Supplier Medchemexpress LLC Supplier No. HY11028410MG
Medchemexpress, HY-110284 10mg BAM 15 CAS:210302-17-3 BAM 15 is a novel mitochondrial protonophore uncoupler. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.