Learn More
Medchemexpress LLC BAM 15 | 210302-17-3 | 99.8% | 340.29 | 10 MG
SDP
Catalog No. 5000343552
Encompass_Preferred
BAM 15 is a mitochondrial protonophore uncoupler and an oxidative phosphorylation (OXPHOS) uncoupler. It increases O2 consumption across a broad dosing range without increasing reactive oxygen species (ROS). This product is for research use only.
  • Increases O2 consumption without increasing ROS
  • Maintains uncoupled respiration at a high rate in various cell lines
  • Induces mitochondrial swelling
  • Protects against kidney injury in animal models
  • More viable cells compared to FCCP-treated cells
Catalog No. 50-003-43552 Supplier Medchemexpress LLC Supplier No. HY11028450MG
BAM 15 is a mitochondrial protonophore uncoupler and an oxidative phosphorylation (OXPHOS) uncoupler. It increases O2 consumption across a broad dosing range without increasing reactive oxygen species (ROS). This product is for research use only.
  • Increases O2 consumption without increasing ROS
  • Maintains uncoupled respiration at a high rate in various cell lines
  • Induces mitochondrial swelling
  • Protects against kidney injury in animal models
  • More viable cells compared to FCCP-treated cells